missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, HPLC Grade, Ricca Chemical
$718.23
Chemical Identifiers
| CAS | 67-64-1 |
|---|---|
| Molecular Formula | C3H6O |
| Molecular Weight (g/mol) | 58.08 |
| MDL Number | MFCD00008765 |
| InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
| Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
| PubChem CID | 180 |
| ChEBI | CHEBI:15347 |
| IUPAC Name | propan-2-one |
| SMILES | CC(C)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
RSOA0011500
|
N/A | 500 mL | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
RSOA00111C
|
N/A | 1 L | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
RSOA00114C
|
N/A | 4 L | Amber Glass Bottle |
Each for $718.23
|
|
||||
Chemical Identifiers
| C3H6O | |
| MFCD00008765 | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CHEBI:15347 | |
| CC(C)=O |
Specifications
| 67-64-1 | |
| MFCD00008765 | |
| CSCPPACGZOOCGX-UHFFFAOYSA-N | |
| propan-2-one | |
| 180 | |
| HPLC |
| C3H6O | |
| acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
| CC(C)=O | |
| 58.08 | |
| CHEBI:15347 | |
| Acetone |