missing translation for 'onlineSavingsMsg'
Learn More

Acetone/Alcohol, 90:10 by Volume, Reagents

Catalog No. 01336019
Change view
Click to view available options
Quantity: 200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01336019 Quantity: 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336019 Supplier Labchem Inc Supplier No. CA015400200E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetone/Alcohol, 90:10 by Volume, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-64-1 , 64-17-5 , 64-17-5
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym Dimethyl Ketone/Reagent Alcohol, 2-Propanone/Reagent Alcohol
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Color Colorless
Quantity 200 L
Packaging Steel Drum
Physical Form Liquid
Specific Gravity 0.789
Boiling Point 56°C
Grade Reagent
Chemical Name or Material Acetone/Alcohol, 90:10 by Volume