Learn More
Acetic Acid, Optima™ LC/MS, Fisher Chemical™
Supplier: Fisher Chemical A11310X1AMP
Specifications
| 0.02 AU max. at 350nm, 0.05 AU max. at 280nm, 1 AU max. at 255nm | |
| 2.5 | |
| 16°C | |
| 117°C | |
| ≥99.7 % | |
| CH3COOH | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| ≥99.7% | |
| 1.048g/cm³ | |
| 40°C | |
| Liquid | |
| 2.10g/mol |
| Acetic Acid | |
| 99.8% | |
| 64-19-7 | |
| Colorless | |
| C2H4O2 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 60.05 | |
| OPTIMA™ LC-MS | |
| 10 x 1 mL | |
| Ampoule | |
| 5ppm max. | |
| 1.52 kPa at 20°C |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:2
Instability:0