missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Optima™ LC/MS, Fisher Chemical™

Catalog No. p-4041825
Change view
Click to view available options
Quantity:
1 mL
50 mL
10 x 1 mL
Packaging:
Ampoule
Nalgene™ Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A1131AMP 1 mL Ampoule
A11310X1AMP 10 x 1 mL Ampoule
A11350 50 mL Nalgene™ Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1131AMP Supplier Thermo Fisher Scientific Supplier No. A113 1AMP
Only null left

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Absorbance 0.02 AU max. at 350nm, 0.05 AU max. at 280nm, 1 AU max. at 255nm
pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Assay Percent Range ≥99.7 %
Linear Formula CH3COOH
Formula Weight 60.05
Percent Purity ≥99.7%
Grade OPTIMA™ LC-MS
Density 1.048g/cm³
Quantity 1 mL
Flash Point 40°C
Packaging Ampoule
Physical Form Liquid
Residue after Evaporation 5ppm max.
Vapor Pressure 1.52 kPa at 20°C
Chemical Name or Material Acetic Acid, 99.8%
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:2
Instability:0