missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid/Iso-Octane, 60/40 v/v Certified to Cd8B-90, Reagents

Catalog No. 01335969
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Poly Bottle
Safety Coated Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01335969 1 L Glass Bottle
01335968 2.5 L Safety Coated Amber Glass Bottle
01335967 4 L Poly Bottle
01335966 4 L Glass Bottle
01335965 4 L Safety Coated Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01335969 Supplier Labchem Inc Supplier No. CA1007001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetic Acid/Iso-Octane, 60/40 v/v Certified to Cd8B-90, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-19-7 , 540-84-1
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Ethanoic Acid/2,2,4-Trimethylpentane
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9065
Grade Reagent
Chemical Name or Material Acetic Acid/Iso-Octane, 60/40 v/v Certified to Cd8B-90