missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial, ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company RABA001025D1
Specifications
| Acetic Acid | |
| 64-19-7 | |
| Colorless | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| ACS | |
| Glass Bottle | |
| max. 0.001% |
| Glacial | |
| 98.0 | |
| 100.0 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 99.7% (min.) | |
| 2.5 L | |
| Liquid |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| CHEBI:15366 | |
| CC(O)=O |
| C2H4O2 | |
| MFCD00036152 | |
| 176 | |
| acetic acid |
Safety and Handling
ShelfLife : 60 months