missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Glacial, ACS Reagent Grade, Ricca Chemical
Specifications
Specifications
| CAS Min % | 98.0 |
| Color | Colorless |
| CAS Max % | 100.0 |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| Percent Purity | 99.7% (min.) |
| Grade | ACS |
| Quantity | 500 mL |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Residue after Evaporation | max. 0.001% |
| Show More |
Safety and Handling
| ShelfLife | 60 months |