missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid Glacial ACS, MilliporeSigma™
Supplier: Technidata AX007375
Specifications
| Acetic Acid | |
| 17°C | |
| 116°C to 118°C (1013hPa) | |
| C2H4O2 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 6.5 | |
| 1.05g/cm3 (20°C) | |
| 4 to 19.9%(V) | |
| 485°C | |
| 15.4 hPa (20°C) |
| Suprapur™ | |
| 64-19-7 | |
| 100% | |
| CH3COOH | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| ACS | |
| 2.5 L | |
| 39.9°C (103.8°F) | |
| Poly Coated Glass Bottle |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Safety and Handling
Recommended Storage : Store at +15°C to 25°C