missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid Glacial ACS, MilliporeSigma™

Catalog No. p-3176262
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Poly Coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MAX00736 500 mL Glass Bottle
MAX0073P5 500 mL Poly Coated Glass Bottle
MAX00739 2.5 L Glass Bottle
MAX007375 2.5 L Poly Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MAX00736 Supplier Technidata Supplier No. AX00736
Only null left

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Melting Point 17°C
Boiling Point 116°C to 118°C (1013hPa)
Assay Percent Range 100%
Linear Formula CH3COOH
Formula Weight 6.5
Grade ACS
Density 1.05g/cm3 (20°C)
Quantity 500 mL
Explosion Limit 4 to 19.9%(V)
Flash Point 39.9°C (103.8°F)
Ignition Point 485°C
Packaging Glass Bottle
Vapor Pressure 15.4 hPa (20°C)
Chemical Name or Material Acetic Acid, Suprapur™
Show More Show Less
Recommended Storage Store at +15°C to 25°C