missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Certified, 6.0N ±0.1N (6.0M), LabChem™

Catalog No. p-5388945
Change view
Click to view available options
Quantity:
1 L
4 L
Catalog No. Quantity
LC104002 1 L
LC104004 4 L
2 options

Catalog No. LC104002

Supplier: LabChem LC104002

Only null left

Chemical Identifiers

CAS 64-19-7 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
CAS Max % 65.42, 34.58
Linear Formula CH3COOH
UN Number UN2790
Solubility Information Soluble in water
Concentration 6.0N ±0.1N (6.0M)
Formula Weight 60.05
Grade Certified
Density 1.04g/mL
Quantity 1 L
Odor Vinegar
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.04g/mL
Traceability to NIST Traceable to NIST
Chemical Name or Material Acetic Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature