missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Certified, 3.0% (v/v) ±0.1%, LabChem™

Catalog No. p-5379572
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC101601 500 mL
LC101604 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC101601 Supplier LabChem Supplier No. LC101601
Only null left

Chemical Identifiers

CAS 64-19-7 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless
CAS Max % 96.87, 3.13
Linear Formula CH3COOH
Solubility Information Soluble in water
Formula Weight 60.05
Percent Purity 3.0% ±0.1% (v/v)
Grade Certified
Density 1g/mL
Quantity 500 mL
Odor Vinegar
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Chemical Name or Material Acetic Acid (3+97)
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin and serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/at
Recommended Storage Room Temperature