missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Certified, 3.0% (v/v) ±0.1%, LabChem™

Supplier:  LabChem LC101604

 View more versions of this product

Catalog No. LC101604

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid
96.87,3.13
C2H4O2
MFCD00036152
Soluble in water
CC(O)=O
60.05
CHEBI:15366
3.0% ±0.1% (v/v)
1g/mL
Vinegar
Liquid
Traceable to NIST
64-19-7,7732-18-5
Colorless
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
60.05
Certified
4 L
Poly Bottle
1g/mL

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin and serious eye irritation.

GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions