missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide acétique, certifié, 1,000N ±0,005N (1,0M), LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Acetic Acid |
| Couleur | Incolore |
| CAS % maximal | 94.04, 5.96 |
| Concentration | 1,000N ±0,005N (1,0M) |
| Densité | 1.01g/mL |
| Quantité | 500 mL |
| Odeur | Vinaigre |
| Emballage | Bouteille en polyéthylène |
| Forme physique | Liquide |
| Gravité spécifique | 1.01g/mL |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin iritation and serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Température ambiante |