Learn More
Acetic Acid, Certified, 1.00% (v/v) ±0.05%, LabChem™
Supplier: LabChem LC101204
Specifications
| Acetic Acid | |
| 99,1 | |
| C2H4O2 | |
| MFCD00036152 | |
| Soluble in water | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| 1.00% ± 0.05% (v/v) | |
| 1g/mL | |
| Vinegar | |
| Liquid | |
| Traceable to NIST |
| 64-19-7 , 7732-18-5 | |
| Colorless | |
| CH3COOH | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 60.05 | |
| Certified | |
| 4 L | |
| Poly Bottle | |
| 1g/mL |
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Safety and Handling
GHS H Statement
Causes skin and serious eye irritation.
GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
Recommended Storage : Room Temperature