missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, Certified, 0.200N ±0.005N (0.2M), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 98.8, 1.2 |
| Concentration | 0.200N ±0.005N (0.2M) |
| Density | 1g/mL |
| Quantity | 1 L |
| Odor | Vinegar |
| Packaging | Poly Bottle |
| Physical Form | Liquid |
| Specific Gravity | 1g/mL |
| Traceability to NIST | Traceable to NIST |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin and serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |