missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Certified, 0.100N ±0.001N (0.1M), LabChem™

$53.07 - $166.63

Chemical Identifiers

CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC103501
View Documents
LabChem
LC103501
500 mL
Each for $53.07
Only null left
 
LC103502
View Documents
LabChem
LC103502
1 L
Each for $88.23
Only null left
 
LC103504
View Documents
LabChem
LC103504
4 L
Each for $166.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

64-19-7,7732-18-5
Colorless
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
60.05
CHEBI:15366
Certified
500 mL
Poly Bottle
1g/mL
Acetic Acid
99.4,0.6
C2H4O2
MFCD00036152
Soluble in water
CC(O)=O
0.100N ±0.001N (0.1M)
176
60.05
1g/mL
Vinegar
Liquid
Traceable to NIST
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'