missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Certified, 0.100N ±0.001N (0.1M), LabChem™

Supplier:  LabChem LC103504

 View more versions of this product

Catalog No. LC103504


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid
99.4,0.6
C2H4O2
MFCD00036152
Soluble in water
CC(O)=O
0.100N ±0.001N (0.1M)
176
60.05
1g/mL
Vinegar
Liquid
Traceable to NIST
64-19-7,7732-18-5
Colorless
CH3COOH
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
60.05
CHEBI:15366
Certified
4 L
Poly Bottle
1g/mL

Chemical Identifiers

64-19-7
60.05
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions