missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 90%, Reagents

Catalog No. 01336066
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336066 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336066 Supplier Labchem Inc Supplier No. CA01220020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetic Acid, 90%, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 64-19-7 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH <2
Boiling Point 118°C
Color Colorless
Quantity 20 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0673
Grade Reagent
Chemical Name or Material Acetic Acid, 90%