missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 9% v/v, Reagents

Catalog No. p-200022058
Change view
Click to view available options
Quantity:
10 L
20 L
55 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335872 10 L
01335871 20 L
01335870 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01335872 Supplier Labchem Inc Supplier No. CA30090010F
Only null left

Acetic Acid, 9% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Percent Purity 9%
Grade Reagent
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0047
Chemical Name or Material Acetic Acid, 9% v/v