missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 56% (w/w), Ricca Chemical
$123.91 - $377.06
Chemical Identifiers
| CAS | 64-19-7 |
|---|---|
| Molecular Formula | C2H4O2 |
| Molecular Weight (g/mol) | Mixture |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| PubChem CID | 176 |
| ChEBI | CHEBI:15366 |
| IUPAC Name | acetic acid |
| SMILES | CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
140516
|
Ricca Chemical Company
140516 |
500 mL | Natural Poly Bottle |
Each for $123.91
|
|
||||
|
140532
|
Ricca Chemical Company
140532 |
1 L | Natural Poly Bottle |
Each for $210.81
|
|
||||
|
14051
|
Ricca Chemical Company
14051 |
4 L | Natural Poly Bottle |
Each for $377.06
|
|
||||
Chemical Identifiers
| 64-19-7 | |
| Mixture | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Specifications
| 64-19-7,7732-18-5 | |
| Colorless | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| acetic acid | |
| 176 | |
| 56% w/w | |
| 500 mL | |
| Liquid |
| 43.0,55.0 | |
| 45.0,57.0 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 56% (w/w) | |
| Mixture | |
| CHEBI:15366 | |
| Laboratory | |
| Natural Poly Bottle | |
| Acetic Acid |