missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 56% (w/w), Ricca Chemical

Supplier:  Ricca Chemical Company 14051

 View more versions of this product

Catalog No. 14051


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid
43.0,55.0
45.0,57.0
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
56% (w/w)
Mixture
CHEBI:15366
Laboratory
Natural Poly Bottle
64-19-7,7732-18-5
Colorless
C2H4O2
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
acetic acid
176
56% w/w
4 L
Liquid

Chemical Identifiers

64-19-7
Mixture
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions