missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 50% v/v, Reagents

Catalog No. 01336635
Change view
Click to view available options
Quantity:
100 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336635 100 mL
01336634 20 L
01336633 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336635 Supplier Labchem Inc Supplier No. C7100610100A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetic Acid, 50% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-19-7 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH <2
Boiling Point 118°C
Color Colorless
Quantity 100 mL
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.0578
Percent Purity 50%
Grade Reagent
Chemical Name or Material Acetic Acid, 50% v/v