missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 50% (v/v), Ricca Chemical

Catalog No. 14016
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
14016 500 mL Amber Glass Bottle
14032 1 L Amber Glass Bottle
1401 4 L Amber Glass Bottle
1405 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14016 Supplier Ricca Chemical Company Supplier No. 14016

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 49.0
Color Colorless
CAS Max % 51.0
Concentration 50% (v/v)
Percent Purity 50% v/v
Grade Laboratory
Quantity 500 mL
Packaging Amber Glass Bottle
Physical Form Liquid
Chemical Name or Material Acetic Acid, 50% (v/v)