missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 5% v/v, Reagents
Specifications
Specifications
| pH | 2.4 |
| Boiling Point | 100°C |
| Color | Colorless |
| Quantity | 10 L |
| Packaging | Cubitainer |
| Physical Form | Liquid |
| Specific Gravity | 1.0073 |
| Percent Purity | 5% |
| Grade | Reagent |
| Chemical Name or Material | Acetic Acid, 5% v/v |