missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 5% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company 13032
Specifications
| Acetic Acid | |
| 7732-18-5,64-19-7 | |
| 96.0,6.0 | |
| Colorless Liquid | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| Mixture | |
| CHEBI:15366 | |
| Laboratory | |
| Natural Poly Bottle |
| 5% (v/v) | |
| 94.0,4.0 | |
| Colorless | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 5% (v/v) | |
| 176 | |
| 4.94 to 5.06% | |
| 1 L | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| CHEBI:15366 | |
| CC(O)=O |
| C2H4O2 | |
| MFCD00036152 | |
| 176 | |
| acetic acid |
Safety and Handling
ShelfLife : 36 months