missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 5% (v/v), Ricca Chemical
$69.59 - $187.69
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | C2H4O2 |
| Molecular Weight (g/mol) | Mixture |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| PubChem CID | 176 |
| ChEBI | CHEBI:15366 |
| IUPAC Name | acetic acid |
| SMILES | CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
130-16
|
Ricca Chemical Company
RI13016 |
500 mL | Natural Poly Bottle |
N/A
|
|
||||
|
R13032
|
Ricca Chemical Company
13032 |
1 L | Natural Poly Bottle |
N/A
|
|
||||
|
130-1
|
Ricca Chemical Company
RI1301 |
4 L | Natural Poly Bottle |
N/A
|
|
||||
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| CHEBI:15366 | |
| CC(O)=O |
| C2H4O2 | |
| MFCD00036152 | |
| 176 | |
| acetic acid |
Specifications
| 7732-18-5,64-19-7 | |
| 96.0,6.0 | |
| Colorless Liquid | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| Mixture | |
| CHEBI:15366 | |
| Laboratory | |
| Natural Poly Bottle | |
| Acetic Acid, 5% (v/v) |
| 94.0,4.0 | |
| Colorless | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 5% (v/v) | |
| 176 | |
| 4.94 to 5.06% | |
| 500 mL | |
| Liquid |
Safety and Handling
ShelfLife : 36 months