missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 4% (v/v), Ricca Chemical

Catalog No. R01250004A
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
Packaging:
Cubitainer
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R01250004A 4 L Natural Poly Bottle
R01250001A 1 L Natural Poly Bottle
R012500010F 10 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R01250004A Supplier Ricca Chemical Company Supplier No. R01250004A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64-19-7 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.052
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(=O)O

Specifications

Assay Percent Range 4.00 ± 0.04% (v/v)
Quantity 4 L
Odor Weak vinegar
Packaging Natural Poly Bottle
Chemical Name or Material Acetic Acid, 4% (v/v) Aqueous Solution