missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 4% (v/v), Ricca Chemical
Specifications
Specifications
| Assay Percent Range | 4.00 ± 0.04% (v/v) |
| Quantity | 1 L |
| Odor | Weak vinegar |
| Packaging | Natural Poly Bottle |
| Chemical Name or Material | Acetic Acid, 4% (v/v) Aqueous Solution |