missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 4% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company R012500010F

Quantity 10 L
Packaging Cubitainer

 View more versions of this product

Catalog No. R012500010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

64-19-7 , 7732-18-5
60.052
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(=O)O
C2H4O2
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid

Specifications

Acetic Acid
64-19-7 , 7732-18-5
C2H4O2
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
176
10 L
Cubitainer
4% (v/v) Aqueous Solution
4.00 ± 0.04% (v/v)
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(=O)O
60.052
CHEBI:15366
Weak vinegar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions