missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 30% (v/v), Ricca Chemical
Specifications
Specifications
| CAS Min % | 75.0, 23.48 |
| Color | Colorless |
| CAS Max % | 78.0, 24.43 |
| Concentration | 30% (v/v) |
| Percent Purity | 30% v/v |
| Grade | Laboratory |
| Quantity | 20 L |
| Packaging | Cubitainer |
| Physical Form | Liquid |
| Chemical Name or Material | Acetic Acid |