missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 30% v/v, Reagents
Acetic Acid, 30% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$122.67 - $1189.33
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Ethanoic Acid, Ethylic Acid |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-335-876
|
Labchem Inc
CA29950010F |
10 L |
Each for $635.83
|
|
|||||
|
01-335-875
|
Labchem Inc
CA2995001A |
1 L |
Each for $122.67
|
|
|||||
|
01-335-874
|
Labchem Inc
CA29950020F |
20 L |
Each for $1,189.33
|
|
|||||
|
01-335-873
|
Labchem Inc
CA2995004A |
4 L |
Each for $305.53
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Ethanoic Acid, Ethylic Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| 2 | |
| 100°C | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 30% | |
| 10 L | |
| Liquid | |
| Acetic Acid, 30% v/v |
| 7732-18-5,64-19-7 | |
| Colorless | |
| Ethanoic Acid, Ethylic Acid | |
| O | |
| 18.02 | |
| Reagent | |
| Cubitainer | |
| 1.0391 |