missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 30% v/v, Reagents
Acetic Acid, 30% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CA29950020F
Specifications
| Acetic Acid, 30% v/v | |
| 7732-18-5,64-19-7 | |
| Colorless | |
| Ethanoic Acid, Ethylic Acid | |
| O | |
| 18.02 | |
| Reagent | |
| Cubitainer | |
| 1.0391 |
| 2 | |
| 100°C | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| 30% | |
| 20 L | |
| Liquid |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Ethanoic Acid, Ethylic Acid | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |