missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 3% v/v, Reagents

Catalog No. 01335916
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335916 1 L
01335915 4 L
01335914 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01335916 Supplier Labchem Inc Supplier No. CA2035001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetic Acid, 3% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name water
SMILES O

Specifications

pH 2.5
Boiling Point 100°C
Color Colorless
Quantity 1 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 1.0045
Percent Purity 3%
Grade Reagent
Chemical Name or Material Acetic Acid, 3% v/v