missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 3% (v/v), Ricca Chemical

Supplier:  Ricca Chemical Company RI1201

 View more versions of this product

Catalog No. R1201


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Acetic Acid
7732-18-5,64-19-7
C2H4O2
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
acetic acid
176
3% v/v
4 L
Liquid
3% (v/v)
Colorless
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
3% (v/v)
Mixture
CHEBI:15366
Laboratory
Natural Poly Bottle

Chemical Identifiers

7732-18-5
Mixture
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions