missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 3% (v/v), Ricca Chemical

$56.09 - $199.13

Chemical Identifiers

CAS 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
120-16
View Documents Promotion Details
Ricca Chemical Company
RI12016
500 mL Natural Poly Bottle
Each for $56.09
Only null left
 
120-32
View Documents Promotion Details
Ricca Chemical Company
12032
1 L Natural Poly Bottle
Each for $77.53
Only null left
 
R1201
View Documents Promotion Details
Ricca Chemical Company
RI1201
4 L Natural Poly Bottle
Each for $199.13
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
Mixture
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

7732-18-5,64-19-7
C2H4O2
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
acetic acid
176
3% v/v
500 mL
Liquid
Colorless
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
3% (v/v)
Mixture
CHEBI:15366
Laboratory
Natural Poly Bottle
Acetic Acid, 3% (v/v)
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'