missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 25% v/v (For AOAC Method 948.10), Fisher Scientific™

Catalog No. SA90251
Change view
Click to view available options
Quantity:
1 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SA90251 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SA90251 Supplier Ricca Chemical Company Supplier No. SA90251

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 74.0, 24.0
CAS Max % 76.0, 26.0
Percent Purity 25% v/v
Grade Laboratory
Quantity 1 L
Packaging Natural Poly Bottle
Chemical Name or Material Acetic Acid, 25% v/v