missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 20% v/v, Reagents

Catalog No. 01335910
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335910 10 L
01335909 1 L
01335908 20 L
01335907 4 L
01335906 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01335910 Supplier Labchem Inc Supplier No. CA20420010F
Only null left

Acetic Acid, 20% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name water
SMILES O

Specifications

pH 2
Boiling Point 118°C
Color Colorless
Percent Purity 20%
Grade Reagent
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0334
Chemical Name or Material Acetic Acid, 20% v/v