missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid (100%, Glacial), Anhydrous, EMSURE™ MilliporeSigma™

Catalog No. M1000632510
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass bottle
Safebreak bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1000632510 2.5 L Safebreak bottle
M1000632500 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1000632510 Supplier Technidata Supplier No. 1.00063.2510

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • 100% anhydrous for analysis

Chemical Identifiers

CAS 64-19-7 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH 2.5 (50g/L, H2O, 20°C)
Melting Point 17°C
Boiling Point 16°C to 118°C (1013hPa)
Color Colorless
Density 1.05g/cm3
Quantity 2.5 L
Explosion Limit 4 to 19.9%
Flash Point 39°C
Ignition Point 485°C
Odor Pungent/Vinegar-like
Packaging Safebreak bottle
Physical Form Liquid
Residue after Evaporation <5ppm
Vapor Pressure 15.4hPa(20°C)
UN Number UN 2789
Solubility Information 602.9g/L
Formula Weight 60.05g/mol
Grade ACS/ISO/Reagent Ph. Eur.
Chemical Name or Material Acetic Acid
Show More Show Less
Hazard Category Corrosive
Hazard Statement H226: Flammable liquid and vapor, H290: May be corrosive to metals, H314: Causes severe skin burns and eye damage,
Precautionary Statement P210: Keep away from heat, P280: Wear protective gloves/ protective clothing/ eye protection/ face protection, P301 + P330 + P331: IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
EINECSNumber 200-580-7
RTECSNumber AF1225000
Recommended Storage Store at 15° to 25°C