missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 10% (v/v), Ricca Chemical

Catalog No. 13532
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Catalog No. Quantity Packaging
13532 1 L Natural Poly Bottle
13516 500 mL Natural Poly Bottle
1351 4 L Natural Poly Bottle
3 options

Catalog No. 13532

Supplier: Ricca Chemical Company 13532

Only null left

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

CAS Min % 89.0, 9.0
Color Colorless
CAS Max % 90.0, 11.0
Appearance Colorless Liquid
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
Concentration 10% (v/v)
Percent Purity 9.94 to 10.06%
Grade Laboratory
Quantity 1 L
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Acetic Acid
Show More Show Less
ShelfLife 36 months