missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 10% v/v, Reagents

Catalog No. 01336102
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336102 10 L
01336101 1 L
01336100 200 L
01336099 20 L
01336098 4 L
01336097 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01336102 Supplier Labchem Inc Supplier No. CA00890010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Acetic Acid, 10% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name water
SMILES O

Specifications

pH 2.3
Boiling Point 118°C
Color Colorless
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0144
Percent Purity 10%
Grade Reagent
Chemical Name or Material Acetic Acid, 10% v/v