missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 1.00 Normal, Reagents

Catalog No. p-200021977
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336113 10 L
01336112 1 L
01336111 20 L
01336110 4 L
01336109 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336113 Supplier Labchem Inc Supplier No. CA00840010F
Only null left

Acetic Acid, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 64-19-7
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Ethanoic Acid, Ethylic Acid
IUPAC Name water
SMILES O

Specifications

pH 2.4
Boiling Point 100°C
Color Colorless
Grade Reagent
Quantity 10 L
Packaging Cubitainer
Physical Form Liquid
Specific Gravity 1.0084
Chemical Name or Material Acetic Acid, 1.00 Normal