missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, 0.100 N (N/10), Ricca Chemical

$73.93 - $196.60

Chemical Identifiers

CAS 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) Mixture
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
145-16
View Documents
Ricca Chemical Company
14516
500 mL Natural Poly Bottle
Each for $73.93
Only null left
 
145-32
View Documents
Ricca Chemical Company
14532
1 L Natural Poly Bottle
Each for $105.23
Only null left
 
145-1
SDS View Documents
Ricca Chemical Company
RI1451
4 L Poly bottle
Each for $196.60
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
Mixture
QTBSBXVTEAMEQO-UHFFFAOYSA-N
176
acetic acid
C2H4O2
MFCD00036152
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CHEBI:15366
CC(O)=O

Specifications

7732-18-5,64-19-7
Colorless
C2H4O2
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
CC(O)=O
acetic acid
176
Laboratory
Natural Poly Bottle
Acetic Acid
97.0,0.59
100.0,0.61
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
0.100 Normal
Mixture
CHEBI:15366
500 mL
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'