missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Acid, 0.100 N (N/10), Ricca Chemical
Supplier: Ricca Chemical Company RI1451
Specifications
| Acetic Acid | |
| 97.0,0.59 | |
| 100.0,0.61 | |
| MFCD00036152 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 0.100 Normal | |
| Mixture | |
| CHEBI:15366 | |
| 4 L | |
| Liquid |
| 7732-18-5,64-19-7 | |
| Colorless | |
| C2H4O2 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| acetic acid | |
| 176 | |
| Laboratory | |
| Poly bottle |
Chemical Identifiers
| 7732-18-5 | |
| Mixture | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |