missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Acetazolamide

Catalog No. 6742100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6742100 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 6742100 Supplier Tocris Bioscience™ Supplier No. 6742/100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Carbonic anhydrase inhibitor

Acetazolamide is a carbonic anhydrase inhibitor. Modulates epileptiform synchronization in rat brain slices. Increases speed of synaptic vesicle recycling and changes synaptic performance in ex vivo murine neuromuscular junction. Lowers intracranial pressure in healthy rats. Also potentiates the antitumor effects of the HDAC inhibitor MS 275 (Cat. No. 6208).

Specifications

Chemical Name or Material N-[5-(Aminosulfonyl)-1,3,4-thiadiazol-2-yl]acetamide
CAS 59-66-5
Quantity 100 mg
Molecular Formula C4H6N4O3S2
Formula Weight 222.25
Purity 99%