missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Buffer, pH 5.5, APHA for Arsenic, Ricca Chemical
Specifications
Specifications
| Color | Clear, Colorless Liquid |
| Concentration | 1X |
| For Use With (Application) | Water Analysis |
| pH | 5.5 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solution Type | Buffer |
| Odor | Acetic acid |
| Packaging | Natural Poly Bottle |
| Sterility | Non-sterile |
| Show More |
Safety and Handling
| Recommended Storage | General |