missing translation for 'onlineSavingsMsg'
Learn More

Acetate Buffer, pH 5.5, APHA for Arsenic, Ricca Chemical

Catalog No. 6316
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6316 500 mL
6332 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6316 Supplier Ricca Chemical Company Supplier No. 6316
Only null left

Chemical Identifiers

CAS 7732-18-5

Specifications

Color Clear, Colorless Liquid
Concentration 1X
For Use With (Application) Water Analysis
pH 5.5
Physical Form Liquid
Quantity 500 mL
Solution Type Buffer
Odor Acetic acid
Packaging Natural Poly Bottle
Sterility Non-sterile
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Chemical Name or Material Acetate Buffer
Show More Show Less
Recommended Storage General