missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetate Buffer, pH 4.0, for Residual Chlorine Analysis, Ricca Chemical
$123.78 - $1315.88
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
50-16
|
Ricca Chemical Company
5016 |
500 mL | Natural Poly Bottle |
Each for $123.78
|
|
||||
|
50-32
|
Ricca Chemical Company
RI5032 |
1 L | Natural Poly Bottle |
Each for $169.03
|
|
||||
|
50-1
|
Ricca Chemical Company
501 |
4 L | Natural Poly Bottle |
Each for $420.22
|
|
||||
|
50-25
|
Ricca Chemical Company
RI5025 |
10 L | Cubitainer |
Each for $819.56
|
|
||||
|
50-5
|
Ricca Chemical Company
RI505 |
20 L | Cubitainer |
Each for $1,315.88
|
|
||||
Chemical Identifiers
| 7732-18-5 |
Specifications
| 7732-18-5 | |
| Water Analysis | |
| Liquid | |
| Buffer | |
| Laboratory |
| 1X | |
| 4 | |
| 500 mL | |
| Natural Poly Bottle | |
| Acetate Buffer |