missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetamiprid

CAS: 135410-20-7 | C10H11ClN4 | 222.68 g/mol

$623.25 - $623.25

Chemical Identifiers

CAS 135410-20-7
Molecular Formula C10H11ClN4
Molecular Weight (g/mol) 222.68
InChI Key WCXDHFDTOYPNIE-UHFFFAOYSA-N
IUPAC Name N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-methylethanimidamide
SMILES CN(CC1=CC=C(Cl)N=C1)C(C)=NC#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC463930010
View Documents
Thermo Scientific Chemicals
463930010
1 g
Each for $623.25
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

135410-20-7
222.68
N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-methylethanimidamide
C10H11ClN4
WCXDHFDTOYPNIE-UHFFFAOYSA-N
CN(CC1=CC=C(Cl)N=C1)C(C)=NC#N

Specifications

135410-20-7
100.0°C to 102.0°C
WCXDHFDTOYPNIE-UHFFFAOYSA-N
N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-methylethanimidamide
1 g
Acetamiprid
C10H11ClN4
Soluble in methanol, chloroform and dimethyl sulfoxide
CN(CC1=CC=C(Cl)N=C1)C(C)=NC#N
222.68
222.67
Videos
Safety and Handling

Safety and Handling

GHS H Statement Toxic if swallowed.

Fatal if inhaled.

Harmful to aquatic life with long lasting effects.

GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

Do not breathe dust/fume/gas/mist/vapors/spray.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Immediately call a POISON CENTER or doctor/physician.

Store in a well-ventilated place.

Keep container tightly closed.

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only