accessibility menu, dialog, popup

UserName

CH3NClNH3CNN(E/Z)

CAS RN 135410-20-7

IUPAC Name: N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-methylethanimidamide
Synonyms: acetamiprid
Molecular Weight (g/mol): 222.68
Molecular Formula: C10H11ClN4
InChi Key: WCXDHFDTOYPNIE-UHFFFAOYSA-N
SMILES: CN(CC1=CC=C(Cl)N=C1)C(C)=NC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results