missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acetamiprid

Catalog No. AC463932500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463932500 250 mg
AC463930010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463932500 Supplier Thermo Scientific Chemicals Supplier No. 463932500

CAS: 135410-20-7 | C10H11ClN4 | 222.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 135410-20-7
Molecular Formula C10H11ClN4
Molecular Weight (g/mol) 222.68
InChI Key WCXDHFDTOYPNIE-UHFFFAOYSA-N
IUPAC Name N-[(6-chloropyridin-3-yl)methyl]-N'-cyano-N-methylethanimidamide
SMILES CN(CC1=CC=C(Cl)N=C1)C(C)=NC#N

Specifications

Melting Point 100.0°C to 102.0°C
Solubility Soluble in methanol, chloroform and dimethyl sulfoxide
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in methanol, chloroform & dimethyl sulfoxi, de
Quantity 250 mg
Formula Weight 222.67
Chemical Name or Material Acetamiprid
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Toxic if swallowed.

Fatal if inhaled.

Harmful to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling.

Do not breathe dust/fume/gas/mist/vapors/spray.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Immediately call a POISON CENTER or doctor/physician.

Store in a well-ventilated place.

Keep container tightly closed.

RUO – Research Use Only